4-tert-Butyl-1-(ethoxycarbonylmethoxy)thiacalix[4]arene ≥94.0% (by HPLC)
Proveedor: TCI
Sinónimos:
4-tert-Butyl-1-(Ethoxycarbonylmethoxy)thiacalix[4]arene, Tetra-tert-butyl[tetrakis(ethoxycarbonylmethoxy)]tetrathiacalix[4]arene
B2310-1G
B2310-5G
TCIAB2310-1GEA
54
EUR
TCIAB2310-1G
TCIAB2310-5G
4-tert-Butyl-1-(ethoxycarbonylmethoxy)thiacalix[4]arene ≥94.0% (by HPLC)
4-tert-Butyl-1-(ethoxycarbonylmethoxy)thiacalix[4]arene
Formula:
C₅₆H₇₂O₁₂S₄ MW: 1065,42 g/mol |
Número MDL:
MFCD06797059 Núm. CAS: 210706-03-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 94.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...