9-Bromononanoic acid ≥97.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
B2323-1G
B2323-5G
TCIAB2323-1GEA
32
EUR
TCIAB2323-1G
TCIAB2323-5G
9-Bromononanoic acid ≥97.0% (by titrimetric analysis)
9-Bromononanoic acid
Formula:
C₉H₁₇BrO₂ Pto. Ebullición: 174°C Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00046564 Núm. CAS: 41059-02-3 UN: 3261 ADR: 8,III |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 35.0 to 39.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...