tert-Butyl-N-(2-hydroxypropyl)carbamate ≥98.0% (by total nitrogen basis)
Proveedor: TCI
Sinónimos:
N-(2-Hydroxypropyl)carbamic Acid tert-Butyl Ester, 1-(tert-Butoxycarbonylamino)-2-propanol, 1-(Boc-amino)-2-propanol, tert-Butyl (2-hydroxypropyl)carbamate
B2418-25G
B2418-5G
TCIAB2418-25GEA
315
EUR
TCIAB2418-25G
TCIAB2418-5G
tert-Butyl-N-(2-hydroxypropyl)carbamate ≥98.0% (by total nitrogen basis)
tert-Butyl-N-(2-hydroxypropyl)carbamate
Formula:
C₈H₁₇NO₃ MW: 175,23 g/mol |
Número MDL:
MFCD02093569 Núm. CAS: 95656-86-3 |
Resultados test especificaciones
Appearance | Colorless to Light yellow clear liquid |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...