Bis(N,N-dimethylacetamide) hydrogen dibromobromate ≥97.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Bis(N,N-dimethylacetamide) Hydrogen Tribromide
B2509-25G
B2509-250G
TCIAB2509-25GEA
50
EUR
TCIAB2509-25G
TCIAB2509-250G
Bis(N,N-dimethylacetamide) hydrogen dibromobromate ≥97.0% (by titrimetric analysis)
Bis(N,N-dimethylacetamide) hydrogen dibromobromate
Formula:
C₈H₁₉Br₃N₂O₂ MW: 414,96 g/mol |
Núm. CAS:
75381-80-5 |
Resultados test especificaciones
Appearance | Light yellow to Brown powder to crystal |
Purity (Iodometric Titration) | min. 97.0 % |
Melting point | 90.0 to 95.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...