Bis(vinylsulphonyl)methane ≥96.0% (por GC)
Proveedor: TCI
|
Atención
|
B2550-25G
B2550-5G
TCIAB2550-25GEA
156
EUR
TCIAB2550-25G
TCIAB2550-5G
Bis(vinylsulphonyl)methane ≥96.0% (por GC)
Bis(vinylsulphonyl)methane
Formula:
C₅H₈O₄S₂ |
Número MDL:
MFCD03265421 Núm. CAS: 3278-22-6 EINECS: 221-909-0 |
Resultados test especificaciones
Appearance | White powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 59.0 to 64.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...