N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,1,2,2-tetramethylethylenediamine ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI

Sinónimos: 6,6'-((1E,1'E)-((2,3-Dimethylbutane-2,3-diyl)bis(azanylylidene))bis(methanylylidene))bis(2,4-di-tert-butylphenol)

B2619-1G B2619-5G
TCIAB2619-1GEA 226 EUR
TCIAB2619-1G TCIAB2619-5G
N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,1,2,2-tetramethylethylenediamine ≥98.0% (by HPLC, total nitrogen)
N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,1,2,2-tetramethylethylenediamine
Formula: C₃₆H₅₆N₂O₂
MW: 548,86 g/mol
Número MDL: MFCD06797077
Núm. CAS: 351498-10-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c893693ea572ab38d06b02bafe182814b296453&vid=3c893693ea572ab38d06b02bafe182814b296453&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 201.0 to 205.0 deg-C
Solubility in THF within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR