2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole ≥99.0% (por GC), purified by sublimation

Proveedor: TCI

Sinónimos: PBD, 2-([1,1-Biphenyl]-4-yl)-5-(4-(tert-butyl)phenyl)-1,3,4-oxadiazole, 2-(4-Biphenylyl)-5-(4-tert-butylphenyl)-1,3,4-oxadiazole

B2696-1G B2696-5G
TCIAB2696-1GEA 45 EUR
TCIAB2696-1G TCIAB2696-5G
2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole ≥99.0% (por GC), purified by sublimation
2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole
Formula: C₂₄H₂₂N₂O
MW: 354,45 g/mol
Número MDL: MFCD00003101
Núm. CAS: 15082-28-7
EINECS: 239-135-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 136.0 to 140.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR