4,6-Bis(diphenylphosphino)phenoxazine ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
Sinónimos:
4,6-Bis(diphenylphosphino)-10H-phenoxazine, NIXANTPHOS
B2717-1G
B2717-100MG
TCIAB2717-1GEA
225
EUR
TCIAB2717-1G
TCIAB2717-100MG
4,6-Bis(diphenylphosphino)phenoxazine ≥98.0% (by HPLC, total nitrogen)
4,6-Bis(diphenylphosphino)phenoxazine
Formula:
C₃₆H₂₇NOP₂ MW: 551,57 g/mol |
Número MDL:
MFCD03788937 Núm. CAS: 261733-18-0 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...