4-Benzoylbutyric acid ≥96.0% (by GC, titration analysis)

Proveedor: TCI
Atención

B2768-25G B2768-5G
TCIAB2768-25GEA 121 EUR
TCIAB2768-25G TCIAB2768-5G
4-Benzoylbutyric acid ≥96.0% (by GC, titration analysis)
4-Benzoylbutyric acid
Formula: C₁₁H₁₂O₃
Número MDL: MFCD00004411
Núm. CAS: 1501-05-9
EINECS: 216-113-5

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 125.0 to 130.0 deg-C

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR