5-Bromo-2-thiophenesulphonamide ≥97.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
5-Bromothiophene-2-sulphonamide
B2793-25G
B2793-5G
TCIAB2793-25GEA
93
EUR
TCIAB2793-25G
TCIAB2793-5G
5-Bromo-2-thiophenesulphonamide ≥97.0% (by GC, titration analysis)
5-Bromo-2-thiophenesulphonamide
Formula:
C₄H₄BrNO₂S₂ MW: 242,11 g/mol |
Número MDL:
MFCD00067990 Núm. CAS: 53595-65-6 |
Resultados test especificaciones
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 139.0 to 143.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...