9-Benzoylcarbazole ≥98.0% (por GC)
Proveedor: TCI
B2864-1G
B2864-5G
TCIAB2864-1GEA
66
EUR
TCIAB2864-1G
TCIAB2864-5G
9-Benzoylcarbazole ≥98.0% (por GC)
9-Benzoylcarbazole
Formula:
C₁₉H₁₃NO |
Número MDL:
MFCD00225468 Núm. CAS: 19264-68-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 99.0 to 103.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...