5-(tert-Butyldiphenylsilyloxy)-1,3-benzenedimethanol ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: [3,5-Bis(hydroxymethyl)phenoxy]-tert-butyldiphenylsilane

B2866-1G B2866-5G
TCIAB2866-1GEA 362 EUR
TCIAB2866-1G TCIAB2866-5G
5-(tert-Butyldiphenylsilyloxy)-1,3-benzenedimethanol ≥98.0% (by HPLC)
5-(tert-Butyldiphenylsilyloxy)-1,3-benzenedimethanol
Formula: C₂₄H₂₈O₃Si
MW: 392.57 g/mol
Núm. CAS: 482627-84-9

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR