Bis(hexylene glycolato)diboron ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
4,4,4',4',6,6'-Hexamethyl-2,2'-bi-1,3,2-dioxaborinane, 4,4,4',4',6,6'-Hexamethyl-2,2'-bi(1,3,2-dioxaborinane)
B2906-1G
B2906-5G
TCIAB2906-1GEA
50
EUR
TCIAB2906-1G
TCIAB2906-5G
Bis(hexylene glycolato)diboron ≥98.0% (por GC)
Bis(hexylene glycolato)diboron
Formula:
C₁₂H₂₄B₂O₄ MW: 253,94 g/mol |
Número MDL:
MFCD06246008 Núm. CAS: 230299-21-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 103.0 to 107.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...