tert-Butyl-3-hydroxyazetidine-1-carboxylate ≥96.0% (por GC)
Proveedor: TCI
|
Atención
|
B2922-1G
B2922-5G
TCIAB2922-1GEA
24
EUR
TCIAB2922-1G
TCIAB2922-5G
tert-Butyl-3-hydroxyazetidine-1-carboxylate ≥96.0% (por GC)
tert-Butyl-3-hydroxyazetidine-1-carboxylate
Formula:
C₈H₁₅NO₃ |
Número MDL:
MFCD04115305 Núm. CAS: 141699-55-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 45.0 to 49.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...