N-(tert-Butoxycarbonyl)-D-glutamic acid ≥97.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: (R)-2-((tert-Butoxycarbonyl)amino)pentanedioic acid, Boc-D-Glu-OH, N-Boc-D-glutamic Acid

B2987-25G B2987-5G
TCIAB2987-25GEA 123 EUR
TCIAB2987-25G TCIAB2987-5G
N-(tert-Butoxycarbonyl)-D-glutamic acid ≥97.0% (by HPLC, titration analysis)
N-(tert-Butoxycarbonyl)-D-glutamic acid
Formula: C₁₀H₁₇NO₆
Número MDL: MFCD00190790
Núm. CAS: 34404-28-9

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Specific rotation [a]20/D +12.0 to +16.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR