N-(tert-Butoxycarbonyl)-D-phenylalanine ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
N-Boc-D-phenylalanine, BOC-D-Phenylalanine, Boc-D-Phe-OH
B2989-25G
B2989-5G
TCIAB2989-25GEA
42
EUR
TCIAB2989-25G
TCIAB2989-5G
N-(tert-Butoxycarbonyl)-D-phenylalanine ≥98.0% (by titrimetric analysis)
N-(tert-Butoxycarbonyl)-D-phenylalanine
Formula:
C₁₄H₁₉NO₄ MW: 265.31 g/mol |
Número MDL:
MFCD00063149 Núm. CAS: 18942-49-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | -22.0 to -28.0 deg (C=1, EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...