(1R,2R)-1,2-Bis(2-hydroxyphenyl)ethylenediamine ≥95.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2,2'-[(1R,2R)-1,2-Diaminoethylene]bisphenol, (1R,2R)-1,2-Diamino-1,2-bis(2-hydroxyphenyl)ethane, 2,2'-((1R,2R)-1,2-Diaminoethane-1,2-diyl)diphenol

B3012-1G B3012-100MG
TCIAB3012-1GEA 724 EUR
TCIAB3012-1G TCIAB3012-100MG
(1R,2R)-1,2-Bis(2-hydroxyphenyl)ethylenediamine ≥95.0% (by HPLC, titration analysis)
(1R,2R)-1,2-Bis(2-hydroxyphenyl)ethylenediamine
Formula: C₁₄H₁₆N₂O₂
MW: 244.29 g/mol
Número MDL: MFCD09751760
Núm. CAS: 870991-70-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a112007e8d2b1be47a280b460189d021a706295f&vid=a112007e8d2b1be47a280b460189d021a706295f&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 95.0 %
Specific rotation [a]20/D 64.0 to 68.0 deg (C=1, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR