Benzo[cd]indol-2(1H)-one ≥97.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Atención
|
B3023-25G
B3023-5G
TCIAB3023-25GEA
174
EUR
TCIAB3023-25G
TCIAB3023-5G
Benzo[cd]indol-2(1H)-one ≥97.0% (by HPLC, total nitrogen)
Benzo[cd]indol-2(1H)-one
Formula:
C₁₁H₇NO |
Número MDL:
MFCD00009748 Núm. CAS: 130-00-7 EINECS: 204-973-4 |
Resultados test especificaciones
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 179.0 to 183.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...