Nε-(tert-butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
B3071-1G
B3071-5G
TCIAB3071-1GEA
26
EUR
TCIAB3071-1G
TCIAB3071-5G
Nε-(tert-butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine ≥98.0% (by HPLC, titration analysis)
Nepsilon-(tert-butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine
Formula:
C₂₆H₃₂N₂O₆ Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00065660 Núm. CAS: 92122-45-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 128.0 to 132.0 deg-C |
Specific rotation [a]20/D | +1.8 to +2.6 deg (C=1, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...