2-Bromo-1,3-difluoro-5-iodobenzene ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
B3073-5G
B3073-25G
TCIAB3073-5GEA
60
EUR
TCIAB3073-5G
TCIAB3073-25G
2-Bromo-1,3-difluoro-5-iodobenzene ≥98.0% (por GC)
2-Bromo-1,3-difluoro-5-iodobenzene
Formula:
C₆H₂BrF₂I MW: 318,89 g/mol |
Número MDL:
MFCD06797752 Núm. CAS: 155906-10-8 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 64.0 to 68.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...