3-Bromo-2-(bromomethyl)propionic acid ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
beta,beta'-Dibromoisobutyric Acid, Bis(bromomethyl)acetic Acid, 3-Bromo-2-(bromomethyl)propanoic acid
B3081-25G
B3081-5G
TCIAB3081-25GEA
247
EUR
TCIAB3081-25G
TCIAB3081-5G
3-Bromo-2-(bromomethyl)propionic acid ≥98.0% (by titrimetric analysis)
3-Bromo-2-(bromomethyl)propionic acid
Formula:
C₄H₆Br₂O₂ MW: 245.90 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00010643 Núm. CAS: 41459-42-1 UN: 3261 ADR: 8 II |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | gt. 98.0 to 103.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...