β-Methasone dipropionate ≥97.0% (by HPLC)

Proveedor: TCI

Sinónimos: Betamethasone 17,21-Dipropionate, 9alpha-Fluoro-11beta,17alpha,21-trihydroxy-16beta-methyl-1,4-pregnadiene-3,20-dione 17,21-Dipropionate

B3166-1G B3166-5G
TCIAB3166-1GEA 61 EUR
TCIAB3166-1G TCIAB3166-5G
β-Methasone dipropionate ≥97.0% (by HPLC)
beta-Methasone dipropionate
Formula: C₂₈H₃₇FO₇
Número MDL: MFCD00133140
Núm. CAS: 5593-20-4
EINECS: 227-005-2
Índice Merck: 14,01180

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 175.0 to 179.0 deg-C
Specific rotation [a]20/D +66.0 to +72.0 deg (C=1, Dioxane)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR