Biotin-PEG2-Amine ≥98.0% (by HPLC)
Proveedor: TCI
Sinónimos:
N-(2-(2-(2-Aminoethoxy)ethoxy)ethyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide, Biotin-DADOO, N-Biotinyl-3,6-dioxaoctane-1,8-diamine, N-[2-[2-(2-Aminoethoxy)ethoxy]ethyl]biotinamide
B3171-25MG
B3171-100MG
TCIAB3171-25MGEA
82
EUR
TCIAB3171-25MG
TCIAB3171-100MG
Biotin-PEG2-Amine ≥98.0% (by HPLC)
Biotin-PEG2-Amine
Formula:
C₁₆H₃₀N₄O₄S MW: 374,50 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD09264745 Núm. CAS: 138529-46-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 13.90 to 16.10 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...