(S,S'')-2,2''-Bis[(S)-4-isopropyloxazolin-2-yl]-1,1''-biferrocene ≥98.0% (by titrimetric analysis)

Proveedor: TCI

B3197-100MG B3197-500MG
TCIAB3197-100MGEA 83 EUR
TCIAB3197-100MG TCIAB3197-500MG
(S,S'')-2,2''-Bis[(S)-4-isopropyloxazolin-2-yl]-1,1''-biferrocene ≥98.0% (by titrimetric analysis)
(S,S'')-2,2''-Bis[(S)-4-isopropyloxazolin-2-yl]-1,1''-biferrocene
Formula: C₃₂H₃₆Fe₂N₂O₂
Núm. CAS: 190601-12-8

Order Now

Resultados test especificaciones

Appearance White to Gray to Brown powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR