4'-(4-Bromophenyl)-2,2':6',2''-terpyridine ≥97.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 4'-(4-Bromophenyl)-alpha,alpha',alpha''-tripyridyl, 4'-(4-Bromophenyl)-2,6':2',2''-terpyridine

B3218-1G B3218-5G
TCIAB3218-1GEA 62 EUR
TCIAB3218-1G TCIAB3218-5G
4'-(4-Bromophenyl)-2,2':6',2''-terpyridine ≥97.0% (by HPLC, titration analysis)
4'-(4-Bromophenyl)-2,2':6',2''-terpyridine
Formula: C₂₁H₁₄BrN₃
MW: 388,27 g/mol
Número MDL: MFCD06796984
Núm. CAS: 89972-76-9

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 160.0 to 164.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR