(R)-N-(tert-Butoxycarbonyl)-2-phenylglycinol ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
Boc-D-Phenylglycinol, (-)-N-Boc-D-α-phenylglycinol
B3270-1G
B3270-5G
TCIAB3270-1GEA
27
EUR
TCIAB3270-1G
TCIAB3270-5G
(R)-N-(tert-Butoxycarbonyl)-2-phenylglycinol ≥98.0% (por GC)
(R)-N-(tert-Butoxycarbonyl)-2-phenylglycinol
Formula:
C₁₃H₁₉NO₃ MW: 237,3 g/mol |
Número MDL:
MFCD00274205 Núm. CAS: 102089-74-7 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 135.0 to 139.0 deg-C |
Specific rotation [a]20/D | -34.0 to -41.0 deg (C=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...