N-(tert-Butoxycarbonyl)-L-2-phenylglycinol ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
Boc-Phenylglycinol, (S)-tert-Butyl (2-hydroxy-1-phenylethyl)carbamate, N-Boc-L-2-phenylglycinol, (S)-2-(tert-Butoxycarbonylamino)-2-phenylethanol
B3272-1G
B3272-5G
TCIAB3272-1GEA
21
EUR
TCIAB3272-1G
TCIAB3272-5G
N-(tert-Butoxycarbonyl)-L-2-phenylglycinol ≥98.0% (por GC)
N-(tert-Butoxycarbonyl)-L-2-phenylglycinol
Formula:
C₁₃H₁₉NO₃ MW: 237,30 g/mol |
Número MDL:
MFCD00274206 Núm. CAS: 117049-14-6 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 134.0 to 139.0 deg-C |
Specific rotation [a]20/D | +32.0 to +39.0 deg (C=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...