2-Butyl-1,3-diazaspiro[4.4]non-1-en-4-one hydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

B3291-25G B3291-5G
TCIAB3291-25GEA 33 EUR
TCIAB3291-25G TCIAB3291-5G
2-Butyl-1,3-diazaspiro[4.4]non-1-en-4-one hydrochloride ≥98.0% (by HPLC, titration analysis)
2-Butyl-1,3-diazaspiro[4.4]non-1-en-4-one hydrochloride
Formula: C₁₁H₁₈N₂O
Número MDL: MFCD03428357
Núm. CAS: 151257-01-1
EINECS: 424-560-4

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR