(+)-Benzotetramisole ≥97.0% (por GC)
Proveedor: TCI
Sinónimos:
(R)-2-Phenyl-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole, (+)-BTM, (2R)-2,3-Dihydro-2-phenylimidazo[2,1-b]benzothiazole
B3296-1G
B3296-200MG
B3296-5G
TCIAB3296-1GEA
134
EUR
TCIAB3296-1G
TCIAB3296-200MG
TCIAB3296-5G
(+)-Benzotetramisole ≥97.0% (por GC)
(+)-Benzotetramisole
Formula:
C₁₅H₁₂N₂S MW: 252,34 g/mol |
Número MDL:
MFCD11112169 Núm. CAS: 885051-07-0 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Optical purity (LC) | min. 98.0 ee% |
Melting point | 91.0 to 95.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...