N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine ≥98.0% (by titrimetric analysis)

Proveedor: TCI

B3303-1G B3303-5G
TCIAB3303-1GEA 56 EUR
TCIAB3303-1G TCIAB3303-5G
N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine ≥98.0% (by titrimetric analysis)
N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine
Formula: C₁₄H₁₈FNO₄
Número MDL: MFCD00079669
Núm. CAS: 57292-45-2

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 98.0 %
Melting point 84.0 to 88.0 deg-C
Specific rotation [a]20/D -17.0 to -23.0 deg (C=1, EtOH)

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR