Boc-Aib-OH ≥98.0% (by GC, titration analysis)
Proveedor: TCI
Sinónimos:
BOC-α-Methylalanine, N-Boc-alpha-methylalanine, 2-(Boc-amino)isobutyric Acid, Boc-Aib-OH
B3323-25G
B3323-5G
TCIAB3323-25GEA
75
EUR
TCIAB3323-25G
TCIAB3323-5G
Boc-Aib-OH ≥98.0% (by GC, titration analysis)
Boc-Aib-OH
Formula:
C₉H₁₇NO₄ MW: 203,24 g/mol |
Número MDL:
MFCD00042973 Núm. CAS: 30992-29-1 EINECS: 250-421-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 118.0 to 122.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...