N6-Benzoyladenine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 6-Benzamidopurine, N-(7H-Purin-6-yl)benzamide

B3344-25G B3344-5G
TCIAB3344-25GEA 227 EUR
TCIAB3344-25G TCIAB3344-5G
N6-Benzoyladenine ≥98.0% (by HPLC, titration analysis)
N6-Benzoyladenine
Formula: C₁₂H₉N₅O
MW: 239,24 g/mol
Número MDL: MFCD00037927
Núm. CAS: 4005-49-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=057902384a64a6bfe0a42fd0d1deb36307cb39e3&vid=cf3f3b5dd643e8a209483c5348f2dabd20d4e506&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 239.0 to 245.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR