Bicyclohexane-4,4'-dione monoethylene ketal ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
B3405-25G
B3405-5G
TCIAB3405-25GEA
109
EUR
TCIAB3405-25G
TCIAB3405-5G
Bicyclohexane-4,4'-dione monoethylene ketal ≥98.0% (por GC)
Bicyclohexane-4,4'-dione monoethylene ketal
Formula:
C₁₄H₂₂O₃ |
Número MDL:
MFCD00809704 Núm. CAS: 56309-94-5 EINECS: 423-860-2 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 100.0 to 104.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...