5-Bromo-6-chloro-3-indolyl-β-D-galactopyranoside ≥98.0% (by HPLC) (contains ca. 10% Ethyl Acetate) for biochemical research

Proveedor: TCI
Peligro

Sinónimos: Magenta-Gal, Mag-Gal

B3469-100MG B3469-20MG
TCIAB3469-100MGEA 140 EUR
TCIAB3469-100MG TCIAB3469-20MG
5-Bromo-6-chloro-3-indolyl-β-D-galactopyranoside ≥98.0% (by HPLC) (contains ca. 10% Ethyl Acetate) for biochemical research
5-Bromo-6-chloro-3-indolyl-beta-D-galactopyranoside
Formula: C₁₄H₁₅BrClNO₆
MW: 408.63 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00210022
Núm. CAS: 93863-88-8

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
beta-Galactosidase detection test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR