4,4'-Bis(hydroxymethyl)-2,2'-bipyridine ≥95.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4,4'-Bis(hydroxymethyl)-2,2'-bipyridyl, [2,2'-Bipyridine]-4,4'-diyldimethanol, 2,2'-Bipyridine-4,4'-dimethanol
B3511-1G
B3511-5G
TCIAB3511-1GEA
126
EUR
TCIAB3511-1G
TCIAB3511-5G
4,4'-Bis(hydroxymethyl)-2,2'-bipyridine ≥95.0% (by GC, titration analysis)
4,4'-Bis(hydroxymethyl)-2,2'-bipyridine
Formula:
C₁₂H₁₂N₂O₂ MW: 216,24 g/mol |
Número MDL:
MFCD06202808 Núm. CAS: 109073-77-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...