Bis(tricyclohexylphosphine)nickel(II) dichloride ≥95.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
Bis(tricyclohexylphosphine)nickel(II) chloride, Dichlorobis(tricyclohexylphosphine)nickel(II)
B3534-1G
B3534-5G
TCIAB3534-1GEA
31
EUR
TCIAB3534-1G
TCIAB3534-5G
Bis(tricyclohexylphosphine)nickel(II) dichloride ≥95.0% (by titrimetric analysis)
Bis(tricyclohexylphosphine)nickel(II) dichloride
Formula:
C₃₆H₆₆Cl₂NiP₂ MW: 690,46 g/mol Pto. de fusión: 227°C |
Número MDL:
MFCD11973802 Núm. CAS: 19999-87-2 |
Resultados test especificaciones
Appearance | Yellow to Amber to Dark red powder to crystal |
Purity (Chelometric Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...