9-Benzylcarbazole ≥98.0% (by GC, total nitrogen)
Proveedor: TCI
Sinónimos:
9-Benzyl-9H-carbazole
B3558-25G
B3558-5G
TCIAB3558-25GEA
372
EUR
TCIAB3558-25G
TCIAB3558-5G
9-Benzylcarbazole ≥98.0% (by GC, total nitrogen)
9-Benzylcarbazole
Formula:
C₁₉H₁₅N MW: 257,34 g/mol Pto. Ebullición: 268°C |
Número MDL:
MFCD00218286 Núm. CAS: 19402-87-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 117.0 to 121.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...