N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
N-Boc-4-chloro-D-phenylalanine, Boc-D-Phe(4-Cl)-OH, (R)-N-BOC-4-Chlorophenylalanine
B3613-1G
B3613-5G
TCIAB3613-1GEA
52
EUR
TCIAB3613-1G
TCIAB3613-5G
N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine ≥98.0% (by HPLC, titration analysis)
N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine
Formula:
C₁₄H₁₈ClNO₄ MW: 299.75 g/mol |
Número MDL:
MFCD00076978 Núm. CAS: 57292-44-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 108.0 to 112.0 deg-C |
Specific rotation [a]20/D | -22.0 to -26.0 deg (C=1, EtOAc) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...