Bis(phenylacetyl)disulphide ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: Phenylacetyl Disulphide, 2-Phenylacetic dithioperoxyanhydride

B3623-5G B3623-25G
TCIAB3623-5GEA 35 EUR
TCIAB3623-5G TCIAB3623-25G
Bis(phenylacetyl)disulphide ≥98.0% (by HPLC)
Bis(phenylacetyl)disulphide
Formula: C₁₆H₁₄O₂S₂
MW: 302,41 g/mol
Número MDL: MFCD00513572
Núm. CAS: 15088-78-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 59.0 to 62.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR