1,3-Di-boc-2-(trifluoromethylsulphonyl)guanidine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

B3625-1G B3625-5G
TCIAB3625-1GEA 150 EUR
TCIAB3625-1G TCIAB3625-5G
1,3-Di-boc-2-(trifluoromethylsulphonyl)guanidine ≥98.0% (by HPLC, titration analysis)
1,3-Di-boc-2-(trifluoromethylsulphonyl)guanidine
Formula: C₁₂H₂₀F₃N₃O₆S
Número MDL: MFCD02683528
Núm. CAS: 207857-15-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR