9,9'-Bifluorenyl ≥98.0% (by HPLC)
Proveedor: TCI
Sinónimos:
9,9'-Bifluorene
B3652-1G
B3652-5G
TCIAB3652-1GEA
55
EUR
TCIAB3652-1G
TCIAB3652-5G
9,9'-Bifluorenyl ≥98.0% (by HPLC)
9,9'-Bifluorenyl
Formula:
C₂₆H₁₈ MW: 330,43 g/mol |
Número MDL:
MFCD00001132 Núm. CAS: 1530-12-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 244.0 to 248.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...