tert-Butyl (4R,6R)-2-[6-[2-[2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetate ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
Sinónimos:
tert-Butyl 2-((4R,6R)-6-(2-(2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)ethyl)-2,2-dimethyl-1,3-dioxan-4-yl)acetate, Atorvastatin Acetonide tert-Butyl Ester
B3690-1G
B3690-5G
TCIAB3690-1GEA
68
EUR
TCIAB3690-1G
TCIAB3690-5G
tert-Butyl (4R,6R)-2-[6-[2-[2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetate ≥98.0% (by HPLC, total nitrogen)
tert-Butyl (4R,6R)-2-[6-[2-[2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetate
Formula:
C₄₀H₄₇FN₂O₅ MW: 654,82 g/mol |
Número MDL:
MFCD04039904 Núm. CAS: 125971-95-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 144.0 to 148.0 deg-C |
Specific rotation [a]20/D | +5.0 to +8.0 deg (C=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...