Cloruro de 4-bromo-2-fluorobencenosulfonilo ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
B3698-5G
B3698-25G
TCIAB3698-5GEA
60
EUR
TCIAB3698-5G
TCIAB3698-25G
Cloruro de 4-bromo-2-fluorobencenosulfonilo ≥98.0% (by GC, titration analysis)
Cloruro de 4-bromo-2-fluorobencenosulfonilo
Formula:
C₆H₃BrClFO₂S MW: 273,51 g/mol |
Número MDL:
MFCD01090993 Núm. CAS: 216159-03-4 UN: 3261 ADR: 8,II |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 63.0 to 67.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...