2-(2H-Benzotriazol-2-yl)-4,6-bis(1-methyl-1-phenylethyl)phenol ≥98.0% (by HPLC)

Proveedor: TCI
Atención

B3717-25G B3717-250G
TCIAB3717-25GEA 17 EUR
TCIAB3717-25G TCIAB3717-250G
2-(2H-Benzotriazol-2-yl)-4,6-bis(1-methyl-1-phenylethyl)phenol ≥98.0% (by HPLC)
2-(2H-Benzotriazol-2-yl)-4,6-bis(1-methyl-1-phenylethyl)phenol
Formula: C₃₀H₂₉N₃O
MW: 447,58 g/mol
Número MDL: MFCD00134705
Núm. CAS: 70321-86-7
EINECS: 274-570-6

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 138.0 to 142.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR