N-(tert-Butoxycarbonyl)-L-tert-leucine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: N-Boc-L-tert-leucine, Boc-Tle-OH, (S)-2-(Boc-amino)-3,3-dimethylbutyric Acid, (S)-2-(tert-Butoxycarbonylamino)-3,3-dimethylbutyric Acid

B3754-5G B3754-25G
TCIAB3754-5GEA 65 EUR
TCIAB3754-5G TCIAB3754-25G
N-(tert-Butoxycarbonyl)-L-tert-leucine ≥98.0% (by HPLC, titration analysis)
N-(tert-Butoxycarbonyl)-L-tert-leucine
Formula: C₁₁H₂₁NO₄
MW: 231,29 g/mol
Número MDL: MFCD00065574
Núm. CAS: 62965-35-9

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 117.0 to 121.0 deg-C
Specific rotation [a]20/D -3.0 to -6.0 deg (C=1、AcOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR