tert-Butyl-2-(hydroxymethyl)piperidine-1-carboxylate ≥97.0% (por GC)
Proveedor: TCI
B3755-25G
B3755-5G
TCIAB3755-25GEA
173
EUR
TCIAB3755-25G
TCIAB3755-5G
tert-Butyl-2-(hydroxymethyl)piperidine-1-carboxylate ≥97.0% (por GC)
tert-Butyl-2-(hydroxymethyl)piperidine-1-carboxylate
Formula:
C₁₁H₂₁NO₃ |
Número MDL:
MFCD04114966 Núm. CAS: 157634-00-9 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 79.0 to 83.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...