N-(tert-Butoxycarbonyl)-L-prolinamide ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
Sinónimos:
tert-Butyl (S)-2-Carbamoyl-1-pyrrolidinecarboxylate, (S)-2-Carbamoyl-1-pyrrolidinecarboxylic Acid tert-Butyl Ester, 1-Boc-L-prolinamide, N-Boc-L-prolinamide, Boc-Pro-NH2
B3756-5G
B3756-1G
TCIAB3756-5GEA
51
EUR
TCIAB3756-5G
TCIAB3756-1G
N-(tert-Butoxycarbonyl)-L-prolinamide ≥98.0% (by HPLC, total nitrogen)
N-(tert-Butoxycarbonyl)-L-prolinamide
Formula:
C₁₀H₁₈N₂O₃ |
Número MDL:
MFCD00190827 Núm. CAS: 35150-07-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 105.0 to 109.0 deg-C |
Specific rotation [a]20/D | -43.0 to -47.0 deg (C=1、MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...