2-Benzyloxy-4-fluorophenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
B3779-1G
B3779-5G
TCIAB3779-1GEA
45
EUR
TCIAB3779-1G
TCIAB3779-5G
2-Benzyloxy-4-fluorophenylboronic acid (contains varying amounts of Anhydride)
2-Benzyloxy-4-fluorophenylboronic acid
Formula:
C₁₃H₁₂BFO₃ |
Número MDL:
MFCD03788402 Núm. CAS: 848779-87-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 108.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...