Nɑ-tert-Butoxycarbonyl-N-γ-trityl-L-asparagine ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
Boc-Asn(Trt)-OH, Nɑ-Boc-Nγ-trityl-L-asparagine
B3789-25G
B3789-5G
TCIAB3789-25GEA
251
EUR
TCIAB3789-25G
TCIAB3789-5G
Nɑ-tert-Butoxycarbonyl-N-γ-trityl-L-asparagine ≥98.0% (by HPLC, titration analysis)
Nalpha-tert-Butoxycarbonyl-N-gamma-trityl-L-asparagine
Formula:
C₂₈H₃₀N₂O₅ MW: 474,56 g/mol Pto. de fusión: 202°C |
Número MDL:
MFCD00153299 Núm. CAS: 132388-68-2 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | -18.0 to -22.0 deg (C=1, methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...