Bentazon ≥97.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

B3825-1G B3825-5G
TCIAB3825-1GEA 79 EUR
TCIAB3825-1G TCIAB3825-5G
Bentazon ≥97.0% (by HPLC, total nitrogen)
Bentazon
Formula: C₁₀H₁₂N₂O₃S
Número MDL: MFCD00078640
Núm. CAS: 25057-89-0
EINECS: 246-585-8
Índice Merck: 14,01053

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 136.0 to 140.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR